Please login to your online account to display your discounted pricing

Bromoform, 99+%, stabilized, ACROS Organics™


Quantity 25mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  220690250

 View more versions of this product

Catalog No. AC220690250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0% (excluding stabilizer) (GC)
Formula Weight 252.73
Physical Form Crystalline Powder
Quantity 25mL
Color White to off-white
Merck Index 15, 1430
Fieser 12,74
Beilstein 01, 68
Density 2.89g/mL
CAS 75-25-2
Assay Percent Range 99+%
Molecular Formula CHBr3
Infrared Spectrum Authentic
Stabilizer 1.5 wt% max. ethanol
CAS 64-17-5
Packaging Glass bottle
Linear Formula CHBr3
Chemical Name or Material Bromoform
Health Hazard 3 GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Toxic if inhaled.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Health Hazard 1 Danger

Specifications

Chemical Name or Material Bromoform
CAS 75-25-2
CAS 64-17-5
Assay ≥99.0% (excluding stabilizer) (GC)
Assay Percent Range 99+%
Molecular Formula CHBr3
Linear Formula CHBr3
Formula Weight 252.73
Accessories

Danger

GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if swallowed.
Toxic if inhaled.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.

SDS & Safety Documents