missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromoferrocene, TCI America™

$142.09 - $482.78

Chemical Identifiers

CAS 1273-73-0
Molecular Formula C10H9BrFe
MDL Number MFCD03093913
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B34761G View Documents TCI America
B34761G
1g Each for $142.09
 
B34765G View Documents TCI America
B34765G
5g Each for $482.78
 
Specifications

Chemical Identifiers

1273-73-0
C10H9BrFe
MFCD03093913

Specifications

Undesignated
1g
C10H9BrFe
264.93
≥95.0% (GC,T)
34°C
1273-73-0
MFCD03093913
Crystal-Powder at 20°C
Bromoferrocene
Safety and Handling

Safety and Handling

TSCA : No