Please login to your online account to display your discounted pricing

Alfa Aesar™ Bromodimethylborane, 97%

Chemical Identifiers

Chemical Name or Material Bromodimethylborane
CAS 5158-50-9
Formula Weight 120.79
MDL Number MFCD00000053
UN Number UN2845
Beilstein 1730828
Synonym Dimethylboron bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3355214

  SDS
alfa aesar™
3355214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 120.79
Chemical Name or Material Bromodimethylborane
Refractive Index 1.399
Physical Form Liquid
Assay 97%
Density 1.238g/mL
MDL Number MFCD00000053
CAS 5158-50-9
UN Number UN2845
Beilstein 1730828
Synonym Dimethylboron bromide
Melting Point −129°C
Boiling Point 31° to 32°C
Quantity 25g
Flash Point −37° (−35°F)
Odor Acrid
Health Hazard 1 Danger
Sensitivity Air & Moisture Sensitive
Health Hazard 2 GHS H Statement
H250-H314-H318
Catches fire spontaneously if exposed to air.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H250-H314-H318
Catches fire spontaneously if exposed to air.
Causes severe skin burns and eye damage.
Causes serious eye damage.