Please login to your online account to display your discounted pricing

Bromodichloromethane, +98%, ACROS Organics™


Quantity 50mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  160700500

 View more versions of this product

Catalog No. AC160700500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4960 to 1.4980 (20°C, 589nm)
Stabilizer potassium carbonate
Density 1.9800g/mL
Color Clear colorless to yellow
Quantity 50mL
Chemical Name or Material Bromodichloromethane
Name Note Stabilized
CAS 75-27-4
Physical Form Clear colorless to yellow liquid
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula CHBrCl2
Linear Formula BrCHCl2
Formula Weight 163.83g/mol
MDL Number MFCD00000824
Beilstein 01, 67
Merck Index 15, 1425
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Suspected of causing genetic defects.
Causes serious eye irritation.
May cause damage to organs.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Bromodichloromethane
Name Note Stabilized
CAS 75-27-4
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula CHBrCl2
Linear Formula BrCHCl2
Formula Weight 163.83g/mol
MDL Number MFCD00000824
Accessories

GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Suspected of causing genetic defects.
Causes serious eye irritation.
May cause damage to organs.

SDS & Safety Documents