missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Bromodeoxyuridine (BrdU) Ab-4, Mouse Monoclonal Antibody

Detect S-phase cells with Thermo Scientific™ Bromodeoxyuridine (BrdU) Ab-4, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

$244.43 - $1,219.86

Specifications

Antigen Bromodeoxyuridine (BrdU) Ab-4
Clone Bu20a
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS1848P Thermo Scientific™ Lab Vision™
MS1848P
Purified Ab with BSA and Azide 1mL Each for $1,122.71
MS1848P0 Thermo Scientific™ Lab Vision™
MS1848P0
Purified Ab with BSA and Azide 100μL Each for $244.43
MS1848P1 Thermo Scientific™ Lab Vision™
MS1848P1
Purified Ab with BSA and Azide 500μL Each for $769.37
MS1848P1ABX Thermo Scientific™ Lab Vision™
MS1848P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $863.51
MS1848PABX Thermo Scientific™ Lab Vision™
MS1848PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,219.86
MS1848R7 Thermo Scientific™ Lab Vision™
MS1848R7
Ready-to-use for IHC (UVLP) 7mL Each for $482.84
Specifications

Description

Detect S-phase cells with Thermo Scientific™ Bromodeoxyuridine (BrdU) Ab-4, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.

Host Species: Mouse

Clone: Bu20a

Isotype: IgG1/κ

Species Reactivity: All species.

Immunogen: BrdU conjugated to BSA

Positive Control: Cultured cells grown in presence of BrdU or tissues from experimental animals injected with BrdU

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Bromodeoxyuridine (BrdU) Ab-4
Immunohistochemistry (Paraffin)
Unlabeled
Murine
Epitope Tags
BrdU conjugated to BSA
Bu20a
Monoclonal
All
RUO
Primary
IgG2a kappa