Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Bromodeoxyuridine (BrdU) Ab-4, Mouse Monoclonal Antibody


Detect S-phase cells with Thermo Scientific™ Bromodeoxyuridine (BrdU) Ab-4, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

Manufacturer: thermo scientific™ lab vision™  MS1848PABX

 View more versions of this product

Catalog No. MS1848PABX

  • / Each
 

Description & Specifications

Specifications

Immunogen BrdU conjugated to BSA
Cross Reactivity All
Regulatory Status RUO
Format Concentrated
Host Species Mouse
Antigen Bromodeoxyuridine (BrdU) Ab-4
Isotype IgG2a kappa
Applications Immunohistochemistry (Paraffin)
Quantity 200µL
Clone Bu20a
Type Primary
Classification Monoclonal
Research Discipline Epitope Tags
Concentration 1mg/mL
Formulation Ab without BSA/Azide (Frozen)
Conjugate Unlabeled

Detect S-phase cells with Thermo Scientific™ Bromodeoxyuridine (BrdU) Ab-4, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.

Host Species: Mouse

Clone: Bu20a

Isotype: IgG1/κ

Species Reactivity: All species.

Immunogen: BrdU conjugated to BSA

Positive Control: Cultured cells grown in presence of BrdU or tissues from experimental animals injected with BrdU

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO