missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Bromodeoxyuridine (BrdU) Ab-4, Mouse Monoclonal Antibody

Detect S-phase cells with Thermo Scientific™ Bromodeoxyuridine (BrdU) Ab-4, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS1848P1ABX

 View more versions of this product

Catalog No. MS1848P1ABX


 

Specifications

Description

Detect S-phase cells with Thermo Scientific™ Bromodeoxyuridine (BrdU) Ab-4, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.

Host Species: Mouse

Clone: Bu20a

Isotype: IgG1/κ

Species Reactivity: All species.

Immunogen: BrdU conjugated to BSA

Positive Control: Cultured cells grown in presence of BrdU or tissues from experimental animals injected with BrdU

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Bromodeoxyuridine (BrdU) Ab-4
Monoclonal
1mg/mL
Ab without BSA/Azide (Frozen)
Murine
IgG2a kappa
RUO
Primary
All
Immunohistochemistry (Paraffin)
Bu20a
Unlabeled
Concentrated
BrdU conjugated to BSA
100μL
Epitope Tags
Monoclonal