Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Bromodeoxyuridine (BrdU) Ab-3, Mouse Monoclonal Antibody

Detect S-phase cells with Thermo Scientific™ Bromodeoxyuridine (BrdU) Ab-3, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

$255.04 - $1,221.76

Specifications

Antigen Bromodeoxyuridine (BrdU) Ab-3
Clone BRD.3
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS1058B

 
thermo scientific™ lab vision™
MS1058B
Biotinylated Ab with BSA and Azide 1mL Each for $1,221.76

MS1058B0

 
thermo scientific™ lab vision™
MS1058B0
Biotinylated Ab with BSA and Azide 100μL Each for $380.22

MS1058B1

 
thermo scientific™ lab vision™
MS1058B1
Biotinylated Ab with BSA and Azide 500μL Each for $915.31

MS1058P

 
thermo scientific™ lab vision™
MS1058P
Purified Ab with BSA and Azide 1mL Each for $882.84

MS1058P0

 
thermo scientific™ lab vision™
MS1058P0
Purified Ab with BSA and Azide 100μL Each for $255.04

MS1058P1

 
thermo scientific™ lab vision™
MS1058P1
Purified Ab with BSA and Azide 500μL Each for $699.98

MS1058P1ABX

 
thermo scientific™ lab vision™
MS1058P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $790.49

MS1058PABX

 
thermo scientific™ lab vision™
MS1058PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,004.82

MS1058R7

 
thermo scientific™ lab vision™
MS1058R7
Ready-to-use for IHC (UVLP) 7mL Each for $504.20
Description & Specifications

Specifications

Antigen Bromodeoxyuridine (BrdU) Ab-3
Clone BRD.3
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Cross Reactivity All
Host Species Murine
Regulatory Status RUO
Research Discipline Epitope Tags
Type Primary
Immunogen Bromodeoxyuridine (BrdU) conjugated to BSA
Isotype IgG1

Detect S-phase cells with Thermo Scientific™ Bromodeoxyuridine (BrdU) Ab-3, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.

Host Species: Mouse

Clone: BRD.3

Isotype: IgG1

Species Reactivity: All species.

Immunogen: Bromodeoxyuridine (BrdU) conjugated to BSA

Positive Control: Cultured cells grown in presence of BrdU or tissues from experimental animals injected with BrdU

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO