Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Bromodeoxyuridine (BrdU) Ab-3, Mouse Monoclonal Antibody


Detect S-phase cells with Thermo Scientific™ Bromodeoxyuridine (BrdU) Ab-3, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

Manufacturer: thermo scientific™ lab vision™  MS1058P1ABX

 View more versions of this product

Catalog No. MS1058P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Regulatory Status RUO
Format Concentrated
Quantity 100µL
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Immunogen Bromodeoxyuridine (BrdU) conjugated to BSA
Cross Reactivity All
Clone BRD.3
Antigen Bromodeoxyuridine (BrdU) Ab-3
Isotype IgG1
Classification Monoclonal
Formulation Ab without BSA/Azide (Frozen)
Conjugate Unlabeled
Type Primary
Research Discipline Epitope Tags
Concentration 1mg/mL
Item Description 100uL; 1mg/mL; without BSA and azide

Detect S-phase cells with Thermo Scientific™ Bromodeoxyuridine (BrdU) Ab-3, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.

Host Species: Mouse

Clone: BRD.3

Isotype: IgG1

Species Reactivity: All species.

Immunogen: Bromodeoxyuridine (BrdU) conjugated to BSA

Positive Control: Cultured cells grown in presence of BrdU or tissues from experimental animals injected with BrdU

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO