Learn More
Bromocyclopropane, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1499618
Description
Bromo Cyclopropane is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. It is also used in the production of cyclopropyl boric acid. It is used as an intermediate in organic synethesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBromo Cyclopropane is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. It is also used in the production of cyclopropyl boric acid. It is used as an intermediate in organic synethesis.
Solubility
It is soluble in chloroform, and methanol. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents, strong bases.
Specifications
Bromocyclopropane | |
1.515 | |
−6°C (21°F) | |
C3H5Br | |
MFCD00001271 | |
UN1993 | |
cyclopropyl bromide, cyclopropane, bromo, cyclopropylbromide, bromo-cyclopropan, bromo-cyclopropane, cyclopropylbrornide, bromanylcyclopropane, pubchem14417, bromocyclopropane, acmc-1aii5 | |
LKXYJYDRLBPHRS-UHFFFAOYSA-N | |
bromocyclopropane | |
78037 | |
99% |
4333-56-6 | |
68°C to 70°C | |
Recognizable | |
1.46 | |
50 g | |
1900287 | |
It is soluble in chloroform,and methanol. Insoluble in water. | |
C1CC1Br | |
120.977 | |
120.98 |
Chemical Identifiers
4333-56-6 | |
120.977 | |
LKXYJYDRLBPHRS-UHFFFAOYSA-N | |
78037 | |
C1CC1Br |
C3H5Br | |
MFCD00001271 | |
cyclopropyl bromide, cyclopropane, bromo, cyclopropylbromide, bromo-cyclopropan, bromo-cyclopropane, cyclopropylbrornide, bromanylcyclopropane, pubchem14417, bromocyclopropane, acmc-1aii5 | |
bromocyclopropane |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H224-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 224-375-7
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only