Learn More
Bromocyclopentane, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1545522
Description
Bromocyclopentane is a precursor used in the preparation of cyclopentene and cyclopentanol. It is a fatty bromide, which is used as a solvent. It is used as an intermediate for the preparation of surfactant and pharmaceuticals and other organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBromocyclopentane is a precursor used in the preparation of cyclopentene and cyclopentanol. It is a fatty bromide, which is used as a solvent. It is used as an intermediate for the preparation of surfactant and pharmaceuticals and other organic compounds.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Bromocyclopentane | |
1.390 | |
42°C (107°F) | |
C5H9Br | |
MFCD00001359 | |
UN1993 | |
cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane | |
BRTFVKHPEHKBQF-UHFFFAOYSA-N | |
bromocyclopentane | |
8728 | |
≥98% |
137-43-9 | |
137°C to 139°C | |
98% | |
1.4895 | |
100 g | |
1209256 | |
Immiscible with water. | |
C1CCC(C1)Br | |
149.031 | |
149.04 | |
Liquid |
Chemical Identifiers
137-43-9 | |
149.031 | |
BRTFVKHPEHKBQF-UHFFFAOYSA-N | |
8728 | |
C1CCC(C1)Br |
C5H9Br | |
MFCD00001359 | |
cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane | |
bromocyclopentane |
Safety and Handling
GHS H Statement
H226
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 205-294-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only