Please login to your online account to display your discounted pricing

Alfa Aesar™ Bromocyclooctane, 97%

$88.36 - $978.99

Chemical Identifiers

Chemical Name or Material Bromocyclooctane
CAS 1556-09-8
Molecular Formula C8H15Br
Formula Weight 191.12
MDL Number MFCD00039603
Beilstein 1901417
Synonym Cyclooctyl bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2483906

  SDS
alfa aesar™
B2483906
5g Each for $88.36

AAB2483922

  SDS
alfa aesar™
B2483922
100g Each for $978.99
Description & Specifications

Specifications

Formula Weight 191.12
Chemical Name or Material Bromocyclooctane
Assay 97%
Molecular Formula C8H15Br
MDL Number MFCD00039603
CAS 1556-09-8
Beilstein 1901417
Synonym Cyclooctyl bromide
Boiling Point 102° to 103°C (18mmHg)
Quantity 5g

Applications
Bromocyclooctane exhibits antibacterial and antineoplastic properties. It is used as pharmaceutical intermediates.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.