missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocyclohexane 98.0+%, TCI America™
Supplier: TCI America B0581500G
Specifications
| Bromocyclohexane | |
| Yellow | |
| C6H11Br | |
| 500 g | |
| AQNQQHJNRPDOQV-UHFFFAOYSA-N | |
| bromocyclohexane | |
| 7960 | |
| ≥98.0% (GC) |
| 108-85-0 | |
| 165°C | |
| MFCD00003819 | |
| cyclohexyl bromide, cyclohexane, bromo, 1-bromocyclohexane, unii-ya0ums4rny, bromo-cyclohexane, ya0ums4rny, cyclohexylbromide, bromocylcohexane, bromocyclohexane, ksc178g7n | |
| BrC1CCCCC1 | |
| 163.06 | |
| 163.06 | |
| Liquid |
Chemical Identifiers
| 108-85-0 | |
| 163.06 | |
| AQNQQHJNRPDOQV-UHFFFAOYSA-N | |
| 7960 | |
| BrC1CCCCC1 |
| C6H11Br | |
| MFCD00003819 | |
| cyclohexyl bromide, cyclohexane, bromo, 1-bromocyclohexane, unii-ya0ums4rny, bromo-cyclohexane, ya0ums4rny, cyclohexylbromide, bromocylcohexane, bromocyclohexane, ksc178g7n | |
| bromocyclohexane |
Safety and Handling
TSCA : Yes