Please login to your online account to display your discounted pricing

Alfa Aesar™ Bromocyclohexane, 99%

$33.76 - $33.76

Chemical Identifiers

Chemical Name or Material Bromocyclohexane
CAS 108-85-0
Molecular Formula C6H11Br
Formula Weight 163.06
MDL Number MFCD00003819
Beilstein 1098373
Merck Index 14,2730
Synonym Cyclohexyl bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3103122

  SDS
alfa aesar™
3103122
100g Each for $33.76

AA3103136

  SDS
alfa aesar
3103136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 163.06
Molecular Formula C6H11Br
Assay Percent Range 99%
CAS 108-85-0
Boiling Point 166° to 167°C
Melting Point −57°C
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Merck Index 14,2730
Chemical Name or Material Bromocyclohexane
Flash Point 62° (143°F)
Sensitivity Air & Light Sensitive
Assay 99%
Synonym Cyclohexyl bromide
Density 1.335g/mL
RTECS Number GU7171700
Health Hazard 1 Warning
TSCA TSCA
Beilstein 1098373
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Quantity 100g
Refractive Index 1.4950
MDL Number MFCD00003819
Physical Form Liquid
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.