Please login to your online account to display your discounted pricing

Bromocresol Purple Sodium Salt, Reagent Grade, LabChem™

$118.43 - $118.43

Chemical Identifiers

Chemical Name or Material Bromocresol Purple Sodium Salt
CAS 62625-30-3
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula C21H15BrNaO5S
Formula Weight 562.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC118857

 
lab chem
LC118857
Amber Glass 10g Each for $118.43
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 562.2
Chemical Name or Material Bromocresol Purple Sodium Salt
Quantity 10g
EINECS Number 263-655-3
Purity Grade Reagent Grade
Linear Formula C21H15BrNaO5S
CAS 62625-30-3
Identification Passes Test
Health Hazard 1 Warning
Packaging Amber Glass
CAS Max % 100
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water
Recommended Storage Room Temperature
Decomposition Information Hydrogen bromide; Carbon monoxide; Carbon dioxide; Sulfur compounds
Melting Point 255°C
Color Pinkish brown
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Room Temperature