missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bromocresol Purple Indicator, Reag. Ph. Eur., Honeywell Fluka™
Indicator, Reag. Ph. Eur.
Supplier: Honeywell-Fluka 326421KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Bromocresol Purple | |
Reag. Ph. Eur. | |
C21H16Br2O5S | |
NONH for all modes of transport | |
ABIUHPWEYMSGSR-UHFFFAOYSA-N | |
2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol | |
8273 | |
540.22g/mol | |
Complies for change range | |
1 kg |
240°C | |
115-40-2 | |
MFCD00011681 | |
359618 | |
CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1 | |
540.22 | |
CHEBI:86154 | |
Glass Bottle | |
5.2 to 6.8 (yellow to purple) |
Chemical Identifiers
115-40-2 | |
540.22 | |
ABIUHPWEYMSGSR-UHFFFAOYSA-N | |
CHEBI:86154 | |
CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1 |
C21H16Br2O5S | |
MFCD00011681 | |
8273 | |
2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol |
Safety and Handling
EINECSNumber : 204-087-8