Please login to your online account to display your discounted pricing

Bromocresol Green Sodium Salt, ACS Grade, LabChem™

$197.69 - $197.69

Chemical Identifiers

Chemical Name or Material Bromocresol Green Sodium Salt
CAS 62625-32-5
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C21H14Br4O5S·Na
Formula Weight 720.06
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC118207

 
lab chem
LC118207
Amber Glass 10g Each for $197.69
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 720.06
CAS 62625-32-5
Packaging Amber Glass
Color Gray
Linear Formula C21H14Br4O5S·Na
CAS Max % 100
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Quantity 10g
Health Hazard 2 GHS H Statement
Product is not hazardous.
Decomposition Information Carbon monoxide; Carbon dioxide; Sulfur compounds; Hydrogen bromide
Solubility Soluble in water
Recommended Storage Room Temperature
Identification Passes Test
Chemical Name or Material Bromocresol Green Sodium Salt
Purity Grade ACS Grade
Melting Point 230°C
EINECS Number 263-657-4
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.

Room Temperature