Please login to your online account to display your discounted pricing

Bromocresol Green-Methyl Red TS, Ricca Chemical


Quantity 500mL
Packaging Poly natural

Manufacturer: ricca chemical company  RI122116

 View more versions of this product

Catalog No. 1221-16

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear red
Quantity 500mL
CAS 7732-18-5
CAS Min % 0.09
CAS 64-17-5
Chemical Name or Material Bromocresol Green-Methyl Red
CAS 7732-18-5 (Water, Deionized)
CAS Max % 0.06
CAS Max % 0.09
CAS Max % 89.0
CAS Max % 12.42
CAS Min % 0.06
Packaging Poly natural
Name Note Test Solution
CAS Min % 11.93
CAS Min % 86.0
CAS 76-60-8
CAS 63451-28-5

Specifications

Chemical Name or Material Bromocresol Green-Methyl Red
Name Note Test Solution
CAS 64-17-5
CAS Min % 86.0
CAS Max % 89.0
CAS 7732-18-5
CAS Min % 11.93
CAS Max % 12.42
CAS 76-60-8
CAS Min % 0.09
CAS Max % 0.09
CAS 63451-28-5
CAS Min % 0.06
CAS Max % 0.06
CAS 7732-18-5 (Water, Deionized)
Physical Form Liquid