missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromocresol Green - Methyl Red Solution (Ethanol Soln.) [mixed Indicator], TCI America™

$55.00 - $128.34

Chemical Identifiers

MDL Number MFCD00134126
Synonym Methyl Red - Bromocresol Green Solution
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B0120100ML View Documents TCI America
B0120100ML
100mL Each for $55.00
 
B0120500ML View Documents TCI America
B0120500ML
500mL Each for $128.34
 
Specifications

Chemical Identifiers

MFCD00134126
Methyl Red - Bromocresol Green Solution

Specifications

MFCD00134126
Methyl Red - Bromocresol Green Solution
Red
Bromocresol Green - Methyl Red Solution (Ethanol Soln.) [mixed Indicator]
1170
Clear Liquid at 20°C
100mL
Safety and Handling

Safety and Handling

TSCA : Yes