Please login to your online account to display your discounted pricing

Bromocresol Green Indicator Aqueous Solution, 1% (w/v), Ricca Chemical


Quantity 1L
Packaging Poly natural

Manufacturer: ricca chemical company  RIR12090001A

 View more versions of this product

Catalog No. R12090001A

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay 1%
Quantity 1L
Physical Form Solution
Assay Percent Range 1% (w/v)
Solubility Solubility in Water: Infinite
Appearance Clear dark blue colored liquid
Odor Odorless
Packaging Poly natural

Specifications

Physical Form Solution
Assay 1%
Assay Percent Range 1% (w/v)