Please login to your online account to display your discounted pricing

Bromocresol Green ACS MP Biomedicals


Quantity 5g

Manufacturer: mp biomedicals  0215250205

 View more versions of this product

Catalog No. ICN15250205

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 5g
Chemical Name or Material Bromocresol green
Name Note Stains and dyes
CAS 76-60-8
Purity Grade ACS Reagent
Physical Form Fine pale orange powder
Molecular Formula C21H14Br4O5S
Formula Weight 698.02g/mol
MDL Number MFCD00005874
Synonym 3', 3''5', 5''-Tetrabromo-m-cresolsulfonephthalein
Recommended Storage Store at Room Temperature (15° to 30°C)
EINECS Number 200-972-8

Specifications

Chemical Name or Material Bromocresol green
Name Note Stains and dyes
CAS 76-60-8
Purity Grade ACS Reagent
Physical Form Fine pale orange powder
Molecular Formula C21H14Br4O5S
Formula Weight 698.02g/mol
MDL Number MFCD00005874
Synonym 3', 3''5', 5''-Tetrabromo-m-cresolsulfonephthalein
Accessories

Store at Room Temperature (15° to 30°C)