Please login to your online account to display your discounted pricing

Bromocresol Green ACS MP Biomedicals

$191.20 - $191.20

Chemical Identifiers

Chemical Name or Material Bromocresol green
Name Note Stains and dyes
CAS 76-60-8
Purity Grade ACS Reagent Grade
Physical Form Fine Powder
Molecular Formula C21H14Br4O5S
Formula Weight 698.02
MDL Number MFCD00005874
Synonym 3', 3''5', 5''-Tetrabromo-m-cresolsulfonephthalein
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15250225

 
mp biomedicals
15250225
25g Each for $191.20
Description & Specifications

Specifications

Purity Grade ACS Reagent Grade
Physical Form Fine Powder
Formula Weight 698.02
Color Pale orange
Quantity 25g
EINECS Number 200-972-8
Name Note Stains and dyes
MDL Number MFCD00005874
CAS 76-60-8
Synonym 3', 3''5', 5''-Tetrabromo-m-cresolsulfonephthalein
Chemical Name or Material Bromocresol green
Molecular Formula C21H14Br4O5S
Recommended Storage Store at Room Temperature (15° to 30°C)