Please login to your online account to display your discounted pricing

Alfa Aesar™ Bromobenzene-d5, 99% (Isotopic)

$145.77 - $145.77

Chemical Identifiers

Chemical Name or Material Bromobenzene-d5
CAS 4165-57-5
Name Note Isotopic
Formula Weight 162.06
MDL Number MFCD00000056
UN Number UN2514
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4226909

 
alfa aesar™
4226909
10g Each for $145.77

AA4226904

 
alfa aesar™
4226904
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.06
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Bromobenzene-d5
Density 1.55g/mL
Name Note Isotopic
Boiling Point 53°C (23mmHg)
Physical Form Liquid
Assay 99%
Quantity 10g
Flash Point 51° (124°F)
Refractive Index 1.557
MDL Number MFCD00000056
CAS 4165-57-5
UN Number UN2514
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315
Flammable liquid and vapor.
Causes skin irritation.
Odor Aromatic
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BROMOBENZENE
Safety and Handling

Warning

GHS P Statement
P210-P280-P303+P361+P353-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315
Flammable liquid and vapor.
Causes skin irritation.