missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromobenzene, 99%

Manufacturer:  Alfa Aesar A1140736

 View more versions of this product

Catalog No. AAA1140736


 

Description

Description

Applications
Bromobenzene is used as solvent in organic synthesis. It is used to prepare the phenylmagnesium bromide, a Grignard reagent, an intermediate in the preparation of benzoic acid by reacting with carbon dioxide. It is a precursor for making phencyclidine. It is useful for the introduction of phenyl group through palladium-catalyzed coupling reactions viz. the Suzuki reaction.

Solubility
Miscible with diethyl ether, alcohol, carbon tetrachloride, chloroform and benzene.

Notes
Store in cool place. Avoid inhalation of vapor or mist. Incompatible with strong oxidizing agents.
Specifications

Specifications

Bromobenzene
1.492g/mL
99%
C6H5Br
UN2514
14,1406
C1=CC=C(C=C1)Br
157.01
CHEBI:3179
108-86-1
500g
99%
MFCD00000055
1236661
QARVLSVVCXYDNA-UHFFFAOYSA-N
bromobenzene
7961
157.02

Chemical Identifiers

108-86-1
157.01
QARVLSVVCXYDNA-UHFFFAOYSA-N
CHEBI:3179
C6H5Br
MFCD00000055
7961
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315

GHS P Statement
P210-P280-P240-P241-P233-P242-P303+P361+P353-P362-P403+P235-P501a

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BROMOBENZENE

RTECSNumber : CY9000000

TSCA : TSCA

SDS Documents