Please login to your online account to display your discounted pricing

Bromoacetyl bromide, 98+%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  106605000

 View more versions of this product

Catalog No. AC106605000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5% (GC)
Physical Form Powder
Formula Weight 201.85
Quantity 500mL
Color White
Beilstein 02, 215
TSCA TSCA
EINECS Number 209-923-5
Packaging Glass bottle
Storage Information May discolor to brown on storage
Infrared Spectrum Authentic
Chemical Name or Material Bromoacetyl bromide
Refractive Index 1.5450 to 1.5490 (20°C, 589nm)
Linear Formula BrCH2COBr
Molecular Formula C2H2Br2O
MDL Number MFCD00000115
Assay Percent Range 98%
Name Note 98+%
CAS 598-21-0
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. Wash face, hands and any exposed skin thoroughly after handling.
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. Reacts violently with water.
Health Hazard 1 Danger

Specifications

Chemical Name or Material Bromoacetyl bromide
Name Note 98+%
CAS 598-21-0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C2H2Br2O
Linear Formula BrCH2COBr
Formula Weight 201.85
MDL Number MFCD00000115
Accessories

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. Wash face, hands and any exposed skin thoroughly after handling.

GHS P Statement Causes severe skin burns and eye damage. Reacts violently with water.

SDS & Safety Documents