missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromoacetyl bromide, 98%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 106601000

 View more versions of this product

Catalog No. AC106601000



Description

Description

Specifications

Specifications

Bromoacetyl bromide
>105°C
Glass bottle
2.31
White
98+%
97.5% min. (GC)
BrCH2COBr
02, 215
Solubility in water: reacts. Other solubilities: solulbe in acetone,carbontetrachloride
C(C(=O)Br)Br
201.85
201.85
98%
2.3100g/mL
Authentic
1.5450 to 1.547
147°C to 150°C
100mL
598-21-0
C2H2Br2O
MFCD00000115
acetyl bromide, 2-bromo, acetyl bromide, bromo, bromacetyl bromide, bromacetylbromide, bromo acetyl bromide, bromoacetic acid bromide, bromoacetyl bromide, bromoacetylbromide, bromoactyl bromide, ccris 9090
LSTRKXWIZZZYAS-UHFFFAOYSA-N
2-bromoacetyl bromide
61147
Powder

Chemical Identifiers

598-21-0
201.85
LSTRKXWIZZZYAS-UHFFFAOYSA-N
61147
C(C(=O)Br)Br
C2H2Br2O
MFCD00000115
acetyl bromide, 2-bromo, acetyl bromide, bromo, bromacetyl bromide, bromacetylbromide, bromo acetyl bromide, bromoacetic acid bromide, bromoacetyl bromide, bromoacetylbromide, bromoactyl bromide, ccris 9090
2-bromoacetyl bromide
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 209-923-5

TSCA : TSCA

Recommended Storage : May discolor to brown on storage

SDS Documents
Product Certifications

RUO – Research Use Only