missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromoacetonitrile, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 221860050

 View more versions of this product

Catalog No. AC221860050


 

Specifications Description & Specifications

Specifications

≥96.0%
119.95
Clear colorless to yellow
13,197
97%
MFCD00001884
Bromoacetonitrile
590-17-0
1.4786 to 1.4806 (20°C, 589nm)
Liquid
5g
02, 216
Authentic
C2H2BrN
97%
BrCH2CN
Glass bottle

Chemical Identifiers

590-17-0
C2H2BrN
119.95
Bromoacetonitrile, 97%
97%
BrCH2CN
MFCD00001884
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement
Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water.

Danger

EINECSNumber : 209-672-1

RTECSNumber : AL7970000