missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromoacetaldehyde Dimethyl Acetal (stabilized with K2CO3), TCI America™

$48.89 - $348.34

Chemical Identifiers

CAS 7252-83-7
Molecular Formula C4H9BrO2
Molecular Weight (g/mol) 169.018
MDL Number MFCD00000213
InChI Key FUSFWUFSEJXMRQ-UHFFFAOYSA-N
Synonym bromoacetaldehyde dimethyl acetal, ethane, 2-bromo-1,1-dimethoxy, 2,2-dimethoxyethyl bromide, dimethylbromoacetal, 1-bromo-2,2-dimethoxyethane, 2,2-dimethoxybromoethane, 2-bromoacetaldehyde dimethyl acetal, bromoacetoaldehyde dimethyl acetal, bromoacetaldehyde dimethylacetal, acetaldehyde, bromo-, dimethyl acetal
PubChem CID 81672
IUPAC Name 2-bromo-1,1-dimethoxyethane
SMILES COC(CBr)OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B109725G View Documents TCI America
B109725G
25g Each for $48.89
 
B1097500G View Documents TCI America
B1097500G
500g Each for $348.34
 
B1097100G View Documents TCI America
B1097100G
100g Each for $143.61
 
Specifications

Chemical Identifiers

7252-83-7
169.018
FUSFWUFSEJXMRQ-UHFFFAOYSA-N
81672
COC(CBr)OC
C4H9BrO2
MFCD00000213
bromoacetaldehyde dimethyl acetal, ethane, 2-bromo-1,1-dimethoxy, 2,2-dimethoxyethyl bromide, dimethylbromoacetal, 1-bromo-2,2-dimethoxyethane, 2,2-dimethoxybromoethane, 2-bromoacetaldehyde dimethyl acetal, bromoacetoaldehyde dimethyl acetal, bromoacetaldehyde dimethylacetal, acetaldehyde, bromo-, dimethyl acetal
2-bromo-1,1-dimethoxyethane

Specifications

150°C
25g
C4H9BrO2
1993
FUSFWUFSEJXMRQ-UHFFFAOYSA-N
2-bromo-1,1-dimethoxyethane
81672
Clear Liquid at 20°C
Bromoacetaldehyde Dimethyl Acetal (stabilized with K2CO3)
Yellow
7252-83-7
MFCD00000213
bromoacetaldehyde dimethyl acetal, ethane, 2-bromo-1,1-dimethoxy, 2,2-dimethoxyethyl bromide, dimethylbromoacetal, 1-bromo-2,2-dimethoxyethane, 2,2-dimethoxybromoethane, 2-bromoacetaldehyde dimethyl acetal, bromoacetoaldehyde dimethyl acetal, bromoacetaldehyde dimethylacetal, acetaldehyde, bromo-, dimethyl acetal
COC(CBr)OC
169.018
169.02
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : Yes