missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromoacetaldehyde dimethyl acetal, 97%, stabilized, Thermo Scientific Chemicals

$224.66 - $224.66

Chemical Identifiers

CAS 7252-83-7
Molecular Formula C4H9BrO2
Molecular Weight (g/mol) 169.02
MDL Number MFCD00000213
InChI Key FUSFWUFSEJXMRQ-UHFFFAOYSA-N
Synonym bromoacetaldehyde dimethyl acetal, ethane, 2-bromo-1,1-dimethoxy, 2,2-dimethoxyethyl bromide, dimethylbromoacetal, 1-bromo-2,2-dimethoxyethane, 2,2-dimethoxybromoethane, 2-bromoacetaldehyde dimethyl acetal, bromoacetoaldehyde dimethyl acetal, bromoacetaldehyde dimethylacetal, acetaldehyde, bromo-, dimethyl acetal
PubChem CID 81672
IUPAC Name 2-bromo-1,1-dimethoxyethane
SMILES COC(CBr)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC221871000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
221871000
100 mL Glass bottle
Each for $224.66
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7252-83-7
169.02
FUSFWUFSEJXMRQ-UHFFFAOYSA-N
81672
COC(CBr)OC
C4H9BrO2
MFCD00000213
bromoacetaldehyde dimethyl acetal, ethane, 2-bromo-1,1-dimethoxy, 2,2-dimethoxyethyl bromide, dimethylbromoacetal, 1-bromo-2,2-dimethoxyethane, 2,2-dimethoxybromoethane, 2-bromoacetaldehyde dimethyl acetal, bromoacetoaldehyde dimethyl acetal, bromoacetaldehyde dimethylacetal, acetaldehyde, bromo-, dimethyl acetal
2-bromo-1,1-dimethoxyethane

Specifications

7252-83-7
148.0°C to 150.0°C
96% min. (GC)
C4H9BrO2
BrCH2CH(OCH3)2
100 mL
1.43
Solubility in water: insoluble
COC(CBr)OC
169.02
169.02
Bromoacetaldehyde dimethyl acetal, 97%
1.4300g/mL
51°C
Glass bottle
1.4450 to 1.4470
MFCD00000213
01, 624
bromoacetaldehyde dimethyl acetal, ethane, 2-bromo-1,1-dimethoxy, 2,2-dimethoxyethyl bromide, dimethylbromoacetal, 1-bromo-2,2-dimethoxyethane, 2,2-dimethoxybromoethane, 2-bromoacetaldehyde dimethyl acetal, bromoacetoaldehyde dimethyl acetal, bromoacetaldehyde dimethylacetal, acetaldehyde, bromo-, dimethyl acetal
FUSFWUFSEJXMRQ-UHFFFAOYSA-N
2-bromo-1,1-dimethoxyethane
81672
97%
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Fatal if inhaled.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat

GHS Signal Word: Danger

EINECSNumber : 230-669-6

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only