missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromoacetaldehyde dimethyl acetal, 97%, stabilized, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 221871000

 View more versions of this product

Catalog No. AC221871000


 

Specifications Description & Specifications

Specifications

Bromoacetaldehyde dimethyl acetal
7252-83-7
97%
BrCH2CH(OCH3)2
MFCD00000213
2-Bromo-1, 1-dimethoxyethane
97%
Glass bottle
C4H9BrO2
169.02
01, 624
100mL

Chemical Identifiers

7252-83-7
C4H9BrO2
169.02
2-Bromo-1, 1-dimethoxyethane
97%
BrCH2CH(OCH3)2
MFCD00000213
Bromoacetaldehyde dimethyl acetal, 97%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Fatal if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 230-669-6

TSCA : TSCA