missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromoacetaldehyde diethyl acetal, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1327636

 View more versions of this product

Catalog No. AAA1327636



Description

Description

Bromoacetaldehyde diethyl acetal, is used as a synthetic building block. It is mainly used for synthesis of antibiotics such as erythromycin and cephalosporins and to other drugs. It also can be used as pharmaceutical intermediates. Besides, this chemical can be used to pruduce drugs such as methylthio imidazole, chlorpheniramine and others for thyroid. It also acts as a Useful intermediate for the preparation of other ?-heterosubstituted acetaldehyde acetals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoacetaldehyde diethyl acetal, is used as a synthetic building block. It is mainly used for synthesis of antibiotics such as erythromycin and cephalosporins and to other drugs. It also can be used as pharmaceutical intermediates. Besides, this chemical can be used to pruduce drugs such as methylthio imidazole, chlorpheniramine and others for thyroid. It also acts as a Useful intermediate for the preparation of other ɑ-heterosubstituted acetaldehyde acetals.

Solubility
It is immiscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Protect from frost, heat and sunlight. Stable under recommended storage conditions. Keep away from Strong oxidizing agents, Acids. Moisture Sensitive.
Specifications

Specifications

Bromoacetaldehyde diethyl acetal
1.27
61°C (142°F)
1.439
MFCD00000214
UN2810
Moisture sensitive
It is immiscible with water.
CCOC(CBr)OCC
197.072
197.08
2032-35-1
66°C to 67°C (18 mmHg)
C6H13BrO2
BrCH2CH(OCH2CH3)2
500 g
635754
bromoacetaldehyde diethyl acetal, bromoacetal, 2,2-diethoxyethyl bromide, ethane, 2-bromo-1,1-diethoxy, bromacetal, 1-bromo-2,2-diethoxyethane, diethyl bromoacetal, 2-bromoacetaldehyde diethyl acetal, 1,1-diethoxy-2-bromoethane, diethyl bromoacetaldehyde acetal
LILXDMFJXYAKMK-UHFFFAOYSA-N
2-bromo-1,1-diethoxyethane
74852
97%

Chemical Identifiers

2032-35-1
197.072
LILXDMFJXYAKMK-UHFFFAOYSA-N
74852
CCOC(CBr)OCC
C6H13BrO2
MFCD00000214
bromoacetaldehyde diethyl acetal, bromoacetal, 2,2-diethoxyethyl bromide, ethane, 2-bromo-1,1-diethoxy, bromacetal, 1-bromo-2,2-diethoxyethane, diethyl bromoacetal, 2-bromoacetaldehyde diethyl acetal, 1,1-diethoxy-2-bromoethane, diethyl bromoacetaldehyde acetal
2-bromo-1,1-diethoxyethane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H227-H315-H319-H335
Toxic if swallowed.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P330-P332+P313-P362-P370+P378q-P501c

H227-H301-H315-H319-H330

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 217-989-1

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only