missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α-Bromo-o-tolunitrile, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC163135000

 View more versions of this product

Catalog No. AC163135000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

α-Bromo-o-tolunitrile
Glass bottle
70°C to 74°C
22115-41-9
100.0
C8H6BrN
MFCD00001794
2 bromomethyl benzonitrile
C1=CC=C(C(=C1)CBr)C#N
196.05
196.05
98%
Authentic
Brown-Gray to White
500g
97.5
97.5% min. (GC)
BrCH2C6H4CN
09, 470
QGXNHCXKWFNKCG-UHFFFAOYSA-N
2-(bromomethyl)benzonitrile
89599
Crystalline Powder or Chunks

Chemical Identifiers

22115-41-9
196.05
QGXNHCXKWFNKCG-UHFFFAOYSA-N
89599
C1=CC=C(C(=C1)CBr)C#N
C8H6BrN
MFCD00001794
2 bromomethyl benzonitrile
2-(bromomethyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents