Please login to your online account to display your discounted pricing

α-Bromo-m-xylene, 96%, ACROS Organics™

$102.96 - $102.96

Chemical Identifiers

Chemical Name or Material α-Bromo-m-xylene
CAS 620-13-3
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C8H9Br
Linear Formula CH3C6H4CH2Br
Formula Weight 185.06
MDL Number MFCD00000177
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC107561000

  SDS
thermo fisher scientific
AC107561000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC107560250

  SDS
thermo fisher scientific
107560250
Glass bottle 25g Each for $102.96
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥95%
Formula Weight 185.06
Quantity 100g
Color Clear colorless to light yellow
Density 1.3700g/mL
Beilstein 05, 374
Merck Index 15, 10288
CAS Min % 95.0
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.5640 to 1.5680 (20°C, 589nm)
Molecular Formula C8H9Br
MDL Number MFCD00000177
CAS 620-13-3
Synonym 3-Methylbenzyl bromide
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material α-Bromo-m-xylene
Assay Percent Range 96%
Linear Formula CH3C6H4CH2Br
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.