Learn More
alpha-Bromo-m-xylene, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 107561000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
α-Bromo-m-xylene | |
95.0 | |
1.3700g/mL | |
82°C | |
96% | |
C8H9Br | |
CH3C6H4CH2Br | |
100g | |
1.37 | |
3-methylbenzyl bromide, 1-bromomethyl-3-methylbenzene, alpha-bromo-m-xylene, m-methylbenzyl bromide, m-xylyl bromide, benzene, 1-bromomethyl-3-methyl, 3-bromomethyl toluene, m-xylene, .alpha.-bromo, 3-methylbenzylbromide, m-xylene, alpha-bromo | |
FWLWTILKTABGKQ-UHFFFAOYSA-N | |
1-(bromomethyl)-3-methylbenzene | |
12099 | |
96% |
620-13-3 | |
100.0 | |
185°C (340.0mmHg) | |
Authentic | |
Glass bottle | |
1.5640 to 1.568 | |
MFCD00000177 | |
05, 374 | |
15, 10288 | |
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol and ether | |
CC1=CC(=CC=C1)CBr | |
185.06 | |
185.06 |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifica
GHS Signal Word: Danger
EINECSNumber : 210-625-2
RUO â Research Use Only