Please login to your online account to display your discounted pricing

α-Bromo-m-xylene, 96%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  107560250

 View more versions of this product

Catalog No. AC107560250

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 185.06
Physical Form Liquid
Assay ≥95%
Quantity 25g
Color Clear colorless to light yellow
Merck Index 15, 10288
Beilstein 05, 374
CAS Min % 95.0
CAS Max % 100.0
Synonym 3-Methylbenzyl bromide
Infrared Spectrum Authentic
CAS 620-13-3
Chemical Name or Material α-Bromo-m-xylene
Molecular Formula C8H9Br
Health Hazard 1 GHS Signal Word: Danger
Refractive Index 1.5640 to 1.5680 (20°C, 589nm)
Packaging Glass bottle
Linear Formula CH3C6H4CH2Br
MDL Number MFCD00000177
Assay Percent Range 96%
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material α-Bromo-m-xylene
CAS 620-13-3
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C8H9Br
Linear Formula CH3C6H4CH2Br
Formula Weight 185.06
MDL Number MFCD00000177
Synonym 3-Methylbenzyl bromide
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

SDS & Safety Documents