missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α-Bromo-m-tolunitrile, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC163510500

 View more versions of this product

Catalog No. AC163510500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

α-Bromo-m-tolunitrile
Glass Bottle
92°C to 97°C
28188-41-2
100.0
C8H6BrN
MFCD00001809
3-bromomethyl benzonitrile
C1=CC(=CC(=C1)CBr)C#N
196.05
196.05
95%
Authentic
Beige to White
50g
94.0
94% min. (GC)
BrCH2C6H4CN
09, II, 326
CVKOOKPNCVYHNY-UHFFFAOYSA-N
3-(bromomethyl)benzonitrile
97249
Crystalline Powder

Chemical Identifiers

28188-41-2
196.05
CVKOOKPNCVYHNY-UHFFFAOYSA-N
97249
C1=CC(=CC(=C1)CBr)C#N
C8H6BrN
MFCD00001809
3-bromomethyl benzonitrile
3-(bromomethyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 248-890-1

SDS Documents