missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3'-nitroacetophenone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 137470250

 View more versions of this product

Catalog No. AC137470250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Bromo-3'-nitroacetophenone
2227-64-7
100.0
Beige-Orange to Green-Yellow
96% min. (GC)
C8H6BrNO3
MFCD00024512
2-bromo-1-3-nitrophenyl ethanone, 2-bromo-3'-nitroacetophenone, 3-nitrophenacyl bromide, 3-nitrophenacylbromide, 2-bromo-1-3-nitrophenyl ethan-1-one, a-bromo-3-nitroacetophenone, 2-bromo-3-nitroacetophenone, alpha-bromo-3'-nitroacetophenone, acetophenone, 2-bromo-3'-nitro, m-nitrophenacyl bromide
GZHPNIQBPGUSSX-UHFFFAOYSA-N
2-bromo-1-(3-nitrophenyl)ethanone
75213
97%
97%
96.0
90°C to 94°C
Authentic
Glass bottle
O2NC6H4COCH2Br
25 g
Solubility in water: insoluble
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)CBr
244.04
244.04
Crystalline Powder

Chemical Identifiers

2227-64-7
244.04
GZHPNIQBPGUSSX-UHFFFAOYSA-N
75213
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)CBr
C8H6BrNO3
MFCD00024512
2-bromo-1-3-nitrophenyl ethanone, 2-bromo-3'-nitroacetophenone, 3-nitrophenacyl bromide, 3-nitrophenacylbromide, 2-bromo-1-3-nitrophenyl ethan-1-one, a-bromo-3-nitroacetophenone, 2-bromo-3-nitroacetophenone, alpha-bromo-3'-nitroacetophenone, acetophenone, 2-bromo-3'-nitro, m-nitrophenacyl bromide
2-bromo-1-(3-nitrophenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 218-764-0

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only