missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dichlorobenzyl bromide, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 146131000

 View more versions of this product

Catalog No. AC146131000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2, 6-Dichlorobenzyl bromide
Authentic
Colorless to White
100g
20443-98-5
100.0
C7H5BrCl2
MFCD00000577
Solubility in water: .. Other solubilities: 1g/10mL methanol
C1=CC(=C(C(=C1)Cl)CBr)Cl
239.93
239.93
99+%
>112°C
Plastic bottle
54°C to 57°C
99+%
99.0
99+%
Cl2C6H3CH2Br
.alpha.-bromo-2,6-dichlorotoluene, 1-bromomethyl-2,6-dichlorobenzene, 2,6 dichloro benzyl bromide, 2,6-dichlorobenzyl bromide, 2,6-dichlorobenzylbromide, 2-bromomethyl-1,3-dichlorobenzene, a-bromo-2,6-dichlorotoluene, alpha-bromo-2,6-dichlorotoluene, benzene, 2-bromomethyl-1,3-dichloro, benzene, 2-bromomethyl-1,3-dichloro-9ci
PDFGFQUSSYSWNI-UHFFFAOYSA-N
2-(bromomethyl)-1,3-dichlorobenzene
30159
Crystals or Crystalline Powder

Chemical Identifiers

20443-98-5
239.93
PDFGFQUSSYSWNI-UHFFFAOYSA-N
30159
C1=CC(=C(C(=C1)Cl)CBr)Cl
C7H5BrCl2
MFCD00000577
.alpha.-bromo-2,6-dichlorotoluene, 1-bromomethyl-2,6-dichlorobenzene, 2,6 dichloro benzyl bromide, 2,6-dichlorobenzyl bromide, 2,6-dichlorobenzylbromide, 2-bromomethyl-1,3-dichlorobenzene, a-bromo-2,6-dichlorotoluene, alpha-bromo-2,6-dichlorotoluene, benzene, 2-bromomethyl-1,3-dichloro, benzene, 2-bromomethyl-1,3-dichloro-9ci
2-(bromomethyl)-1,3-dichlorobenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 243-827-4

RTECSNumber : XS7966000

TSCA : TSCA

SDS Documents