Please login to your online account to display your discounted pricing

α-Bromo-2,4-difluorotoluene, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material α-Bromo-2, 4-difluorotoluene
CAS 23915-07-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H5BrF2
Linear Formula F2C6H3CH2Br
Formula Weight 207.02
MDL Number MFCD00011649
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC303580050

  SDS
thermo fisher scientific
303580050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 207.02
Quantity 5g
Color Clear yellow
CAS Max % 100.0
Chemical Name or Material α-Bromo-2, 4-difluorotoluene
Refractive Index 1.5240 to 1.5260 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C7H5BrF2
Linear Formula F2C6H3CH2Br
Infrared Spectrum Authentic
Packaging Glass bottle
MDL Number MFCD00011649
CAS 23915-07-3
Synonym 2, 4-Difluorobenzyl bromide
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.