missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α-Bromo-2,4-difluorotoluene, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 303580050

 View more versions of this product

Catalog No. AC303580050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

α-Bromo-2, 4-difluorotoluene
97.5
Authentic
1.5240 to 1.5260 (20°C, 589nm)
≥97.5%
C7H5BrF2
207.02
Clear yellow
5g
23915-07-3
100.0
Glass bottle
Liquid
98%
F2C6H3CH2Br
MFCD00011649
2, 4-Difluorobenzyl bromide

Chemical Identifiers

23915-07-3
100.0
C7H5BrF2
207.02
2, 4-Difluorobenzyl bromide
≥97.5%
97.5
98%
F2C6H3CH2Br
MFCD00011649
α-Bromo-2, 4-difluorotoluene

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger