Please login to your online account to display your discounted pricing

Bromine Water, Saturated, Certified, ≥0.20N, LabChem™

$69.66 - $69.66

Chemical Identifiers

Chemical Name or Material Bromine Water
CAS 7732-18-5
CAS Max % 98.7
CAS 7726-95-6
CAS Max % 1.3
Name Note Saturated
Purity Grade Certified Grade
Linear Formula Br
Formula Weight 79.904
UN Number UN1744
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC120001

 
lab chem
LC120001
Amber Glass 500mL Each for $69.66
Description & Specifications

Specifications

Formula Weight 79.904
Purity Grade Certified Grade
Packaging Amber Glass
Concentration ≥0.20N
Quantity 500mL
Health Hazard 3 GHS P Statement
Do not breathe vapors.
Wear protective gloves, eye protection, protective clothing, face protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Health Hazard 1 Danger
CAS Max % 98.7
CAS 7732-18-5
Recommended Storage Room Temperature
Color Clear, red-brown
Density 1g/mL
Identification Passes Test
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life.
Chemical Name or Material Bromine Water
UN Number UN1744
Traceability to NIST Traceable to NIST
Specific Gravity 1g/mL
Solubility Information Soluble in water
Physical Form Liquid
Linear Formula Br
CAS Max % 1.3
CAS 7726-95-6
Name Note Saturated
Safety and Handling

Danger

GHS P Statement
Do not breathe vapors.
Wear protective gloves, eye protection, protective clothing, face protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life.

Room Temperature