missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bromine Water, Saturated, approximately 3% (w/v), Ricca Chemical
Manufacturer: Ricca Chemical Company 119516
Specifications
Bromine Water | |
7726-95-6,7732-18-5 | |
99.0,3.07 | |
brom, brome, bromine, bromine solution, bromine water, bromo, bromo italian, bromo spanish, broom, dibromine | |
BrBr | |
159.808 | |
CHEBI:29224 | |
Safety-coated Glass Bottle | |
500 mL |
Saturated | |
95.0,2.95 | |
Br2 | |
GDTBXPJZTBHREO-UHFFFAOYSA-N | |
molecular bromine | |
24408 | |
Liquid | |
Brown-Red to Orange | |
approximately 3% (w/v) |
Chemical Identifiers
7726-95-6 | |
159.808 | |
brom, brome, bromine, bromine solution, bromine water, bromo, bromo italian, bromo spanish, broom, dibromine | |
CHEBI:29224 | |
BrBr |
Br2 | |
GDTBXPJZTBHREO-UHFFFAOYSA-N | |
24408 | |
molecular bromine |