Please login to your online account to display your discounted pricing

Bromine liquid, ACS, 99.5%, Alfa Aesar™

$284.78 - $284.78

Chemical Identifiers

Chemical Name or Material Bromine liquid
CAS 7726-95-6
Purity Grade ACS Grade
MDL Number MFCD00010896
UN Number UN1744
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA3322036

  SDS
alfa aesar
3322036*
Each for $284.78
Description & Specifications

Specifications

Purity Grade ACS Grade
Quantity 500g
Chemical Name or Material Bromine liquid
Odor Unpleasant
TSCA TSCA
Physical Form Liquid
Assay 99.5%
MDL Number MFCD00010896
CAS 7726-95-6
UN Number UN1744
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H330-H314;
Fatal if inhaled.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: BROMINE
Color Red-brown
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H330-H314;
Fatal if inhaled.
Causes severe skin burns and eye damage.