missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromine liquid, 99.998% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 87614AD

 View more versions of this product

Catalog No. AA87614AD



Specifications

Specifications

Bromine liquid
99.998% (metals basis)
MFCD00010896
GDTBXPJZTBHREO-UHFFFAOYSA-N
molecular bromine
24408
Liquid
Unpleasant
Red to Brown
7726-95-6
Br2
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
BrBr
159.808
CHEBI:29224
3.12g/cm3 at 20°C
173mm Hg at 20°C
50mL

Chemical Identifiers

7726-95-6
159.808
GDTBXPJZTBHREO-UHFFFAOYSA-N
24408
molecular bromine
Br2
MFCD00010896
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
CHEBI:29224
BrBr
Safety and Handling

Safety and Handling

H330-H314-H400: Fatal if inhaled. Causes severe skin burns and eye damage. Very toxic to aquatic life.

DOTInformation : DOT Class: 8, Packaging Group: I

EINECSNumber : 231-778-1

SDS Documents