missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromine liquid, 99.998% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 87614AA

 View more versions of this product

Catalog No. AA87614AA



Specifications

Specifications

Bromine liquid
99.998% (metals basis)
MFCD00010896
GDTBXPJZTBHREO-UHFFFAOYSA-N
molecular bromine
24408
Liquid
173mm Hg at 20°C
5mL
7726-95-6
Br2
bromine or bromine solutions un1744 corrosive, bromine solution 1.0 m in trimethyl phosphate, bromine trace metals basis, bromine puriss. p.a rt, bromine jis special grade, epa pesticide chemical code 008701, bromine saj first grade, bromine acs reagent, bromine or bromine solutions
BrBr
159.808
CHEBI:29224
Unpleasant
Red to Brown

Chemical Identifiers

7726-95-6
159.808
GDTBXPJZTBHREO-UHFFFAOYSA-N
24408
molecular bromine
Br2
MFCD00010896
bromine or bromine solutions un1744 corrosive, bromine solution 1.0 m in trimethyl phosphate, bromine trace metals basis, bromine puriss. p.a rt, bromine jis special grade, epa pesticide chemical code 008701, bromine saj first grade, bromine acs reagent, bromine or bromine solutions
CHEBI:29224
BrBr
Safety and Handling

Safety and Handling

H330-H314-H400: Fatal if inhaled. Causes severe skin burns and eye damage. Very toxic to aquatic life.

DOTInformation : DOT Class: 8, Packaging Group: I

EINECSNumber : 231-778-1

SDS Documents