missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromine concentrate, For 1L standard solution, 0.05 M Br2, Honeywell Fluka™

for 1L standard solution, 0.05 M Br2

Manufacturer:  Honeywell-Fluka 380406X1EA

 View more versions of this product

Catalog No. 60046680



Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Bromine concentrate
7726-95-6,7758-01-2,7758-01-2
≤90.0000%,≤30.0000%
Br2
MFCD00003445
GDTBXPJZTBHREO-UHFFFAOYSA-N
molecular bromine
24408
Concentrate
1.307 g/mL at 25 °C(lit.)
For 1L standard solution, 0.05 M Br2
≥70.0000%,≥10.0000%
6 x 1 ea.
BaCl2
brom, brome, bromine, bromine solution, bromine water, bromo, bromo italian, bromo spanish, broom, dibromine
BrBr
159.808
CHEBI:29224
Ampoule

Chemical Identifiers

7726-95-6
159.808
GDTBXPJZTBHREO-UHFFFAOYSA-N
24408
molecular bromine
Br2
MFCD00003445
brom, brome, bromine, bromine solution, bromine water, bromo, bromo italian, bromo spanish, broom, dibromine
CHEBI:29224
BrBr
Safety and Handling

Safety and Handling

ShelfLife : 1080 days from date of manufacture

Recommended Storage : Room Temp

SDS
Documents

Documents

Product Certifications
Promotions

Promotions