Please login to your online account to display your discounted pricing

Bromine (Certified ACS), Fisher Chemical


Quantity 250mL
Packaging Amber Glass

Manufacturer: fisher scientific company  B385250

 View more versions of this product

Catalog No. B385-250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.5%
Purity Grade Certified ACS Grade
Formula Weight 159.81
Melting Point −7.2°C
Boiling Point 58.7°C
Quantity 250mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Color Red Brown
Density 3.111g/cm³
Atomic Weight 79.91
Vapor Density 5.51g/mol
Vapor Pressure 230mbar at 20°C
Viscosity 0.314 cs at 25°C
Sulfur Compounds 0.001% max.
Residue after Evaporation 0.005% max.
Assay Percent Range ≥99.5 %
Chlorine (Cl) 0.05% max.
Heavy Metals (as Pb) 2ppm max.
ChemAlert Storage Symbol White
MDL Number MFCD00010896
Nickel (Ni) 5ppm max.
Identification Pass Test
Packaging Amber Glass
Iodine (I) 0.001% max.
Cleanup Symbol Solvent
CAS 7726-95-6
Chemical Name or Material Bromine
Molecular Formula Br2

Specifications

Chemical Name or Material Bromine
CAS 7726-95-6
Purity Grade Certified ACS Grade
Assay ≥99.5%
Assay Percent Range ≥99.5 %
Molecular Formula Br2
Formula Weight 159.81
MDL Number MFCD00010896
Accessories
   

SDS & Safety Documents