Please login to your online account to display your discounted pricing

Bromine (Certified ACS), Fisher Chemical

$1,049.92 - $1,127.36

Chemical Identifiers

Chemical Name or Material Bromine
CAS 7726-95-6
Purity Grade Certified ACS Grade
Assay ≥99.5%
Assay Percent Range ≥99.5%
Molecular Formula Br2
Formula Weight 159.81
MDL Number MFCD00010896
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

B385-50

  SDS
fisher scientific company
FLB38550
Amber Glass 50mL Each for $1,049.92

B385-250

  SDS
fisher scientific company
B385250
Amber Glass 250mL Each for $1,127.36
Description & Specifications

Specifications

Assay ≥99.5%
Purity Grade Certified ACS Grade
Melting Point −7.2°C
Boiling Point 58.7°C
Quantity 50mL
Formula Weight 159.81
Assay Percent Range ≥99.5%
UN Number 1744
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Color Red Brown
Density 3.111g/cm³
Vapor Pressure 230mbar at 20°C
Viscosity 0.314 cs at 25°C
Atomic Weight 79.91
Residue after Evaporation 0.005% max.
Sulfur Compounds 0.001% max.
Packaging Amber Glass
Cleanup Symbol Solvent
ChemAlert Storage Symbol White
MDL Number MFCD00010896
Nickel (Ni) 5ppm max.
Chlorine (Cl) 0.05% max.
Identification Pass Test
Heavy Metals (as Pb) 2ppm max.
Chemical Name or Material Bromine
Iodine (I) 0.001% max.
CAS 7726-95-6
Molecular Formula Br2
Safety and Handling