Learn More
Bromine, 99.5+%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 445875000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
58.7°C | |
-7.2°C | |
7726-95-6 | |
99.5% min. (Iodometry) | |
Br2 | |
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish | |
CPELXLSAUQHCOX-UHFFFAOYSA-M | |
molecular bromine | |
24408 | |
159.82 | |
ACS Reagent | |
3.111 |
Bromine | |
1M Solution in trimethyl Phosphate | |
500 g | |
Br | |
15,1401 | |
Solubility in water: 35g/L (20°C). Other solubilities: soluble in concd hcl,aq solns of bromides,freely soluble in alcohol,ether,chcl3,ccl4,cs2 | |
[Br-] | |
79.91 | |
CHEBI:29224 | |
≥99.5% | |
Plastic Coated Glass Bottle | |
3.1110g/mL |
Chemical Identifiers
7726-95-6 | |
79.91 | |
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish | |
CHEBI:29224 | |
[Br-] |
Br | |
CPELXLSAUQHCOX-UHFFFAOYSA-M | |
24408 | |
molecular bromine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 231-778-1
RUO – Research Use Only