Please login to your online account to display your discounted pricing

Bromine-1,4-dioxane complex, 0.25M solution in 1,4-dioxane, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  388418000

 View more versions of this product

Catalog No. AC388418000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Concentration 0.2 to 0.3M
Packaging AcroSeal™ Glass Bottle
Color Clear red
Quantity 800mL
Chemical Name or Material Bromine-1, 4-dioxane complex
Name Note 0.25M solution in 1, 4-dioxane
CAS 15481-39-7
CAS 123-91-1
Physical Form Clear red liquid
Molecular Formula C4H8Br2O2
Formula Weight 247.91g/mol
MDL Number MFCD00034818
Synonym Dioxane dibromide
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
Fatal if inhaled.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repea
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Bromine-1, 4-dioxane complex
Name Note 0.25M solution in 1, 4-dioxane
CAS 15481-39-7
CAS 123-91-1
Molecular Formula C4H8Br2O2
Formula Weight 247.91g/mol
MDL Number MFCD00034818
Synonym Dioxane dibromide
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing cancer.
Fatal if inhaled.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repea

SDS & Safety Documents